Order Entry
Belgium
ContactUsLinkComponent
Anti-Claudin 3 Mouse Monoclonal Antibody [clone: ABT-CLD3]
Anti-Claudin 3 Mouse Monoclonal Antibody [clone: ABT-CLD3]
Fournisseur:  ANTIBODIES.COM
New Product
Certificats
undefined
Anti-Claudin 3 Mouse Monoclonal Antibody [clone: ABT-CLD3]
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Catalog No:
  • ANTIA97074-100
  • ANTIA97074-50
  • Symbole de l'antigène:
  • Claudin 3
  • Claudin 3
  • Nom de l'antigène:
  • C7orf1
  • C7orf1
  • Synonymes antigène:
  • Claudin3|Claudin 3|CPE receptor 2|Cldn3|Claudin-3|CPE R2|CLD3_HUMAN|CLDN 3|Clostridium perfringens enterotoxin receptor 2
  • Claudin3|Claudin 3|CPE receptor 2|Cldn3|Claudin-3|CPE R2|CLD3_HUMAN|CLDN 3|Clostridium perfringens enterotoxin receptor 2
  • Type d'anticorps:
  • Primaire
  • Primaire
  • Clonalité:
  • Monoclonal
  • Monoclonal
  • Conjugaison:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-CLD3
  • ABT-CLD3
  • Réactivité:
  • Human
  • Human
  • Hôte:
  • Mouse
  • Mouse
  • ID de gène:
  • UniprotID# O15551
  • UniprotID# O15551
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChimie:
  • Yes
  • Yes
  • Immunogène:
  • Synthetic peptide derived from human Claudin 3
  • Synthetic peptide derived from human Claudin 3
  • Formulaire:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Température de transport:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC
  • IHC

Spécifications

Frequently Bought Together