Anti-PKC delta Rabbit Monoclonal Antibody [clone: ARC1434]
N° de catalogue ANTIA308143-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CVID9
- Symbole de l'antigène:PKC delta
- Clonalité:Monoclonal
- Clone:ARC1434
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q05655
- Synonymes antigène:Kinase PKC delta|KPCD|nPKC-delta|D14Ertd420e|45047|KPCD_HUMAN|nPKC delta|MGC49908
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:78 kDa
- Sequence:PFHGDDEDELFESIRVDTPHYPRWITKESKDILEKLFEREPTKRLGVTGNIKIHPFFKTINWTLLEKRRLEPPFRPKVKSPRDYSNFDQEFLNEKARLSYSDKNLID
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 547-653 of human PKC delta (Q05655)
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Type de conditionnement:Plastic vial
- Cdt:100 µl
Spécifications
À propos de cet article
Rabbit monoclonal [ARC1434] antibody to PKC delta for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: PKC delta
Clonality: Monoclonal
Clone: ARC1434
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat