Anti-ALDH7A1 Rabbit Monoclonal Antibody [clone: ARC2837]
Catalog # ANTIA308004-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:26g turgor protein homolog
- Symbole de l'antigène:ALDH7A1
- Clonalité:Monoclonal
- Clone:ARC2837
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P49419
- Synonymes antigène:Aldehyde dehydrogenase 7 family, member A1|Alpha-AASA dehydrogenase|Alpha AASA dehydrogenase|AL7A1_HUMAN|Aldehyde dehydrogenase family 7 member A1|Aldehyde dehydrogenase 7 A1|ALDH7A1|Alpha-aminoadipic semialdehyde dehydrogenase|Alpha aminoadipic semialdehyde dehydrogenase
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:58 kDa
- Sequence:MWRLPRALCVHAAKTSKLSGPWSRPAAFMSTLLINQPQYAWLKELGLREENEGVYNGSWGGRGEVITTYCPANNEPIARVRQASVADYEETVKKAREAWK
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ALDH7A1 (P49419)
- Purification:Affinity purification
- Type de conditionnement:Plastic vial
- Cdt:100 µl
Specifications
About this item
Rabbit monoclonal [ARC2837] antibody to ALDH7A1 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: ALDH7A1
Clonality: Monoclonal
Clone: ARC2837
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse