Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Proteasome 20S LMP7 Rabbit Polyclonal Antibody
  ANTIA306731-100
New Product
undefined
Anti-Proteasome 20S LMP7 Rabbit Polyclonal Antibody
  ANTIA306731-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    ALDD
  • Symbole de l'antigène:
    Proteasome 20S LMP7
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P28062
  • Synonymes antigène:
    D6S216E|Low molecular mass protein 7|Macropain subunit C13|LMP 7|Large multifunctional protease 7|LMP7|Low molecular weight protein 7|Large multifunctional peptidase 7|D6S216
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    23 kDa
  • Sequence:
    LSMGSMICGWDKKGPGLYYVDEHGTRLSGNMFSTGSGNTYAYGVMDSGYRPNLSPEEAYDLGRRAIAYATHRDSYSGGVVNMYHMKEDGWVKVESTDVSDLLHQYREANQ
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant fusion protein containing a sequence corresponding to amino acids 163-272 of human PSMB8 (NP_004150.1).
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µl

 

 

Rabbit polyclonal antibody to Proteasome 20S LMP7 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Proteasome 20S LMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat