Order Entry
Belgium
ContactUsLinkComponent
Anti-CACNA1C Goat Polyclonal Antibody
Anti-CACNA1C Goat Polyclonal Antibody
N° de catalogue ANTIA83471-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-CACNA1C Goat Polyclonal Antibody
N° de catalogue ANTIA83471-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    alpha-1 polypeptide
  • Symbole de l'antigène:
    CACNA1C
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • Synonymes antigène:
    cardiac muscle|CACN 2|isoform 1|L type|CACN2|CACH 2|CAC1C_HUMAN|CACNA1C|CACH2
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
  • Sequence:
    C-RARGRPSEEELQD
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Synthetic peptide corresponding to Human CACNA1C (C terminal)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Spécifications

À propos de cet article

Goat polyclonal antibody to CACNA1C for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, WB

Type: Primary
Antigen: CACNA1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human