Anti-Doublecortin Goat Polyclonal Antibody
ANTIA83146-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:DBCN
- Symbole de l'antigène:Doublecortin
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:Dbct|DCX_HUMAN|Doublin|Doublecortex|Lis X|Doublecortin|FLJ51296|DCX|DC
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-KTSANMKAPQS
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide corresponding to Human DCX (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Goat polyclonal antibody to Doublecortin for ELISA, WB, IHC and IF with samples derived from human, mouse and rat.
- Validated applications: ELISA, WB, IHC, IF
Type: Primary
Antigen: Doublecortin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat