Anti-160 kD Neurofilament Medium Goat Polyclonal Antibody
Catalog # ANTIA82582-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:150kDa medium
- Symbole de l'antigène:160 kD Neurofilament Medium
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:Neurofilament 3|NEF3|NFM|160 kD Neurofilament Medium|NF160|NEFM
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-QASHITVERKDYLK
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide corresponding to Human NEFM (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Goat polyclonal antibody to 160 kD Neurofilament Medium for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: 160 kD Neurofilament Medium
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse