Anti-VEGFA Recombinant Recombinant Antibody [clone: DMC276]
Fournisseur: ANTIBODIES.COM
New Product
Certificats
À propos de cet article
Recombinant chimeric monoclonal [DMC276] antibody to VEGFA for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: VEGFA
Clonality: Recombinant
Clone: DMC276
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Spécifications
- Catalog No:
- ANTIA318782-500
- ANTIA318782-100
- Symbole de l'antigène:
- VEGFA
- VEGFA
- Nom de l'antigène:
- Folliculostellate cell-derived growth factor
- Folliculostellate cell-derived growth factor
- Synonymes antigène:
- vascular endothelial growth factor|MVCD1|vascular endothelial growth factor A165|MGC70609|Vegf|Vascular permeability factor|Glioma-derived endothelial cell mitogen|Vascular endothelial growth factor A|vascular endothelial growth factor A121
- vascular endothelial growth factor|MVCD1|vascular endothelial growth factor A165|MGC70609|Vegf|Vascular permeability factor|Glioma-derived endothelial cell mitogen|Vascular endothelial growth factor A|vascular endothelial growth factor A121
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Recombinant
- Recombinant
- Conjugaison:
- Unconjugated
- Unconjugated
- Clone:
- DMC276
- DMC276
- Réactivité:
- Human
- Human
- Hôte:
- Recombinant
- Recombinant
- ID de gène:
- UniprotID# P15692
- UniprotID# P15692
- Isotype:
- IgG1
- IgG1
- Cytométrie en flux:
- Yes
- Yes
- Formulaire:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Température de stockage:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Température de transport:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Spécifications
Frequently Bought Together