Order Entry
Belgium
ContactUsLinkComponent
Anti-Apolipoprotein B Mouse Monoclonal Antibody [clone: APOB/4333]
Anti-Apolipoprotein B Mouse Monoclonal Antibody [clone: APOB/4333]
  ANTIA252095-100
New Product
undefined
Anti-Apolipoprotein B Mouse Monoclonal Antibody [clone: APOB/4333]
  ANTIA252095-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Apo B-100
  • Symbole de l'antigène:
    Apolipoprotein B
  • Clonalité:
    Monoclonal
  • Clone:
    APOB/4333
  • Conjugaison:
    Unconjugated
  • Hôte:
    Mouse
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG1 kappa
  • Réactivité:
    Human
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P04114
  • Synonymes antigène:
    Apolipoprotein B-48|Apolipoprotein B (including Ag(x) antigen)|Apo B-48|ApoB 48|Apolipoprotein B|APOB|Apolipoprotein B 100|APOB_HUMAN|ApoB 100
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    512 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant fragment, around amino acids 592 - 689, of human APOB protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Mouse monoclonal [APOB/4333] antibody to Apolipoprotein B for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Apolipoprotein B
Clonality: Monoclonal
Clone: APOB/4333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human