Anti-FBXO22 Rabbit Polyclonal Antibody
ANTIA10406-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:F box only protein 22
- Symbole de l'antigène:FBXO22
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q8NEZ5
- Synonymes antigène:1600016C16Rik|F-box only protein 22|F box protein FBX22p44|FBX22|FBXO 22|F box protein 22|FBX22_HUMAN|F-box protein FBX22p44|0610033L19Rik
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:45 kDa
- Sequence:YMADSETFISLEECRGHKRARKRTSMETALALEKLFPKQCQVLGIVTPGIVVTPMGSGSNRPQEIEIGESGFALLFPQIEGIKIQPFHFIKDPKNLTLERHQLTEVGLLDNPELRVVLVFGYNCCKVGASNYLQQVVSTFSDMNIILAGGQVDNLSSLTSEKNPLDIDASGVVGLSFSGHRIQSATVLLNEDVSDEKTAEAAMQRLKAANIPEHNTIGFMFACVGRGFQYYRAKGNVEADAFRKFFPSVPLFGFFGNGEIGCDRIVTGNFILRKCNEVKDDDLFHSYTTIMALIHLGSSK
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 104 - 403 of human FBXO22 (NP_671717.1)
- Purification:Affinity purification
- Type de conditionnement:Plastic vial
- Cdt:100 µl
Rabbit polyclonal antibody to FBXO22 for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: FBXO22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human