Order Entry
Belgium
ContactUsLinkComponent
Anti-AARS2 Rabbit Polyclonal Antibody
Anti-AARS2 Rabbit Polyclonal Antibody
N° de catalogue ANTIA10232-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-AARS2 Rabbit Polyclonal Antibody
N° de catalogue ANTIA10232-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Alanine tRNA ligase 2 mitochondrial
  • Symbole de l'antigène:
    AARS2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# Q5JTZ9
  • Synonymes antigène:
    AlaRS|AARSL|AARS 2|Alanine tRNA ligase|Alanyl tRNA synthetase 2 mitochondrial|bA444E17.1|Alanyl-tRNA synthetase 2|AARS2|Alanyl tRNA synthetase like
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    107 kDa
  • Sequence:
    RRELLATVKMLQRRANTAIRKLQMGQAAKKTQELLERHSKGPLIVDTVSAESLSVLVKVVRQLCEQAPSTSVLLLSPQPMGKVLCACQVAQGAMPTFTAEAWALAVCSHMGGKAWGSRVVAQGTGSTTDLEAALSIAQTYALSQL
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant fusion protein containing a sequence corresponding to amino acids 841 - 985 of human AARS2 (NP_065796.1)
  • Purification:
    Affinity purification
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µl

Spécifications

À propos de cet article

Rabbit polyclonal antibody to AARS2 for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: AARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human