Order Entry
Belgium
ContactUsLinkComponent
 
Anti-ERp57 Goat Polyclonal Antibody
New Product
 
undefined
Anti-ERp57 Goat Polyclonal Antibody
New Product

 

  • Catalog No:
  • ANTIA121548-15
  • ANTIA121548-600
  • Symbole de l'antigène:
  • ERp57
  • ERp57
  • Nom de l'antigène:
  • 58 kDa glucose regulated protein
  • 58 kDa glucose regulated protein
  • Synonymes antigène:
  • Disulfide isomerase ER 60|Disulfide isomerase ER-60|ER protein 57|Endoplasmic reticulum resident protein 60|58 kDa glucose-regulated protein|58 kDa microsomal protein|Endoplasmic reticulum resident protein 57|ER p57|ER protein 60
  • Disulfide isomerase ER 60|Disulfide isomerase ER-60|ER protein 57|Endoplasmic reticulum resident protein 60|58 kDa glucose-regulated protein|58 kDa microsomal protein|Endoplasmic reticulum resident protein 57|ER p57|ER protein 60
  • Type d'anticorps:
  • Primaire
  • Primaire
  • Clonalité:
  • Polyclonal
  • Polyclonal
  • Conjugaison:
  • Unconjugated
  • Unconjugated
  • Réactivité:
  • Monkey, Human, Dog, Rat, Mouse
  • Monkey, Human, Dog, Rat, Mouse
  • Hôte:
  • Goat
  • Goat
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChimie:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogène:
  • Recombinant peptide derived from within residues a.a. 300 to the C terminus of human ERp57, expressed in and purified from E. coli
  • Recombinant peptide derived from within residues a.a. 300 to the C terminus of human ERp57, expressed in and purified from E. coli
  • Formulaire:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purification
  • Affinity purification
  • Concentration:
  • 3 mg/ml
  • 3 mg/ml
  • Storage buffer:
  • Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
  • Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
  • Température de stockage:
  • Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
  • Température de transport:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC-P, IHC-Fr
  • IHC-P, IHC-Fr

 

Frequently Bought Together