Anti-Cytokeratin 15 Rabbit Recombinant Antibody [clone: KRT15/8312R]
Catalog # ANTIA316241-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:AI528832
- Symbole de l'antigène:Cytokeratin 15
- Clonalité:Recombinant
- Clone:KRT15/8312R
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG kappa
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P19012
- Synonymes antigène:Ka15|Cytokeratin 15|CK 15|K1C15_HUMAN|K1CO|Cytokeratin15|CK15|CK-15|Cytokeratin-15
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:52 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Température de transport:Shipped on blue ice at +4 °C
- Immunogène:Recombinant human full-length KRT15 protein.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRT15/8312R] antibody to cytokeratin 15 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Cytokeratin 15
Clonality: Recombinant
Clone: KRT15/8312R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human