Anti-NUT Rabbit Recombinant Antibody [clone: SNUPN/7351R]
ANTIA315417-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:NUT
- Clonalité:Recombinant
- Clone:SNUPN/7351R
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG kappa
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q86Y26
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:120 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fragment (around Amino Acids 900-1100) of human NUTM1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Recombinant rabbit monoclonal [SNUPN/7351R] antibody to NUT for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: NUT
Clonality: Recombinant
Clone: SNUPN/7351R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human