Anti-HLA Class 1 ABC Rabbit Recombinant Antibody [clone: MHC-I/8147R]
N° de catalogue ANTIA315085-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:HLA A
- Symbole de l'antigène:HLA Class 1 ABC
- Clonalité:Recombinant
- Clone:MHC-I/8147R
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG kappa
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P01889, P30443, P30499
- Synonymes antigène:HLA class 1 B|MHC class I HLA B|HLA Class 1 ABC|HLA class 1 A|HLA B|HLA class 1 C|MHC class I HLA A|Major histocompatibility complex, class I, A + B + C|HLA C
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:40 kDa/41 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fragment (around Amino Acids 100-300) of human HLA-I protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Spécifications
À propos de cet article
Recombinant rabbit monoclonal [MHC-I/8147R] antibody to HLA Class 1 ABC for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HLA Class 1 ABC
Clonality: Recombinant
Clone: MHC-I/8147R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human