Order Entry
Belgium
ContactUsLinkComponent
 
Anti-TCF25 Mouse Monoclonal Antibody [clone: PCRP-TCF25-1A11]
  ANTIA248541-100
New Product
undefined
Anti-TCF25 Mouse Monoclonal Antibody [clone: PCRP-TCF25-1A11]
  ANTIA248541-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    FKSG26
  • Symbole de l'antigène:
    TCF25
  • Clonalité:
    Monoclonal
  • Clone:
    PCRP-TCF25-1A11
  • Conjugaison:
    Unconjugated
  • Cytométrie en flux:
    Yes
  • Hôte:
    Mouse
  • Isotype:
    IgG2a
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# Q9BQ70
  • Synonymes antigène:
    Transcription factor 25|Hulp1|NULP1|PRO2620|hKIAA1049|TCF25_HUMAN|Tcf25|Nuclear localized protein 1|TCF-25
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    77 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant full-length human TCF25 protein
  • Purification:
    Protein A/G chromatography
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Mouse monoclonal [PCRP-TCF25-1A11] antibody to TCF25 for flow cytometry and WB with samples derived from human.

  • Validated applications: Flow cytometry, WB
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml

Type: Primary
Antigen: TCF25
Clonality: Monoclonal
Clone: PCRP-TCF25-1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human