- Cdt:96 Tests
- Durée du test:Multiple steps
- Type de test:Sandwich
- Format:Pre-coated
- Hôte:
- Primary antibody reactivity:Mouse
- Cible:Complement C1q
- Désignation:Mouse complement C1q ELISA kit
- Environmentally Preferable:
- Type déchantillon:Serum, plasma, tissue homogenates, and other biological fluids
- Réactivité croisée:Mouse Complement C1q ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Complement C1q. No significant cross-reactivity or interference between Complement C1q and analogues was observed.
- Méthode de détection:Colorimetric
- Time to Results:4 h 30 min
- Principe du test:Quantitative
- Durée de conservation:Store for 6 months at 4 °C
- Portée de détection:31,25 - 2000 pg/ml
- Température de stockage:4 °C
- Sensibilité:18,75 pg/ml
- Regulatory status:RUO
Mouse Complement C1q ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement C1q in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection Range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Mouse Complement C1q ELISA kit (A303547) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Complement C1q in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C1q has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C1q present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Complement C1q antibody, which binds the captured Complement C1q present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C1q captured in each well. The concentration of Complement C1q can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.