Order Entry
Belgium
ContactUsLinkComponent
Human E-Selectin/CD62E ELISA Kit
Human E-Selectin/CD62E ELISA Kit
N° de catalogue ANTIA78046-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  
undefined
Human E-Selectin/CD62E ELISA Kit
N° de catalogue ANTIA78046-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  

Spécifications

  • Cdt:
    96 Tests
  • Durée du test:
    Multiple steps
  • Type de test:
    Sandwich
  • Format:
    Pre-coated
  • Hôte:
  • Primary antibody reactivity:
    Human
  • Cible:
    E-Selectin/CD62E
  • Désignation:
    Human E-selectin/CD62E ELISA kit
  • Environmentally Preferable:
  • Type déchantillon:
    Serum, plasma, tissue homogenates and other biological fluids
  • Réactivité croisée:
    Human E-Selectin/CD62E ELISA kit exhibits high specificity and excellent specificity for the detection of human E-Selectin/CD62E. No significant cross-reactivity or interference between E-Selectin/CD62E and analogues was observed.
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Principe du test:
    Quantitative
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    78,125 - 5000 pg/ml
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    100 μl
  • Sensibilité:
    46,875 pg/ml
  • Regulatory status:
    RUO

Spécifications

À propos de cet article

Human E-Selectin/CD62E ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human E-Selectin/CD62E in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%

Human E-Selectin/CD62E ELISA kit (A78046) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human E-Selectin/CD62E in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for E-Selectin/CD62E has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the E-Selectin/CD62E present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-E-Selectin/CD62E Antibody, which binds the captured E-Selectin/CD62E present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of E-Selectin/CD62E captured in each well. The concentration of E-Selectin/CD62E can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.