Anti-CPSF4L Rabbit Polyclonal Antibody
N° de catalogue PRSI55-946
Fournisseur: ProSci Inc.
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Putative cleavage and polyadenylation specificity factor subunit 4-like protein
- Symbole de l'antigène:CPSF4L
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:Amino Acid Range = 150-178
- Formulaire:Liquid
- ID de gène:A6NMK7
- Synonymes antigène:CPSF4L
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:21 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Température de transport:4 °C
- Immunogène:This CPSF4L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 150-178 amino acids from the C-terminal region of human CPSF4L.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Taille:400 µl
- Cdt:400 µl
Spécifications
À propos de cet article
The specific function of the protein remains unknown.
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: CPSF4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human