Anti-ALKBH1 Rabbit Polyclonal Antibody
PRSI15-361
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:AlkB homolog 1, histone H2A dioxygenase
- Symbole de l'antigène:ALKBH1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:8846
- Synonymes antigène:DNA demethylase ALKBH1|ABH|alkylation repair homolog 1|hABH|tRNA N1-methyl adenine demethylase|DNA oxidative demethylase ALKBH1|alkylated DNA repair protein alkB homolog 1|DNA 6mA demethylase|DNA N6-methyl adenine demethylase|alkB|nucleic acid dioxygenase ALKBH1|DNA lyase ABH1|ABH1|alkB homolog|alpha-ketoglutarate-dependent dioxygenase ABH1|alkylation repair|ALKBH
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 110-389 of human ALKBH1 (NP_006011.2)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: ALKBH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human