Anti-CYP2W1 Rabbit Polyclonal Antibody
PRSI15-189
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Cytochrome P450 family 2 subfamily W member 1
- Symbole de l'antigène:CYP2W1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:54905
- Synonymes antigène:cytochrome P450 2W1|CYPIIW1|cytochrome P45family 2|polypeptide 1|subfamily W
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 141-490 of human CYP2W1 (NP_060251.2)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: CYP2W1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse