Anti-GGA2 Rabbit Polyclonal Antibody
PRSI14-174
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:golgi-associated, gamma adaptin ear containing, ARF binding protein 2
- Symbole de l'antigène:GGA2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:23062
- Synonymes antigène:VEAR|GGA2|KIAA1080
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GGA2 (NP_055859.1)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: GGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat