Anti-NSD2 Rabbit Polyclonal Antibody
PRSI23-292
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Wolf-Hirschhorn syndrome candidate 1
- Symbole de l'antigène:NSD2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:7468
- Synonymes antigène:MMSET|NSD2|WHS|TRX5|WHSC1|REIIBP
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human WHSC1 (NP_579877.1)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: NSD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse