Anti-ASNS Rabbit Polyclonal Antibody
PRSI16-388
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Asparagine synthetase [glutamine-hydrolyzing]
- Symbole de l'antigène:ASNS
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:440
- Synonymes antigène:ASNS|Glutamine-dependent asparagine synthetase|TS11|Asparagine synthetase [glutamine-hydrolyzing]|Cell cycle control protein TS11
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 262-561 of human ASNS (NP_597680,2)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: ASNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat