Anti-RAPH1 Rabbit Polyclonal Antibody
PRSI16-081
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
- Symbole de l'antigène:RAPH1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:65059
- Synonymes antigène:proline-rich EVH1 ligand 2|RalGDS/AF-6|PREL-2|LPD|lamellipodin|candidate 9|ALS2CR9|amyotrophic lateral sclerosis 2 (juvenile) chromosome region|PREL2|ras-associated and pleckstrin homology domains-containing protein 1|RMO1|candidate 18|ALS2CR18
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human RAPH1 (NP_998754,1)
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: RAPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse