Anti-UGT1A4 Rabbit Polyclonal Antibody
PRSI19-725
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:UDP glucuronosyltransferase 1 family, polypeptide A4
- Symbole de l'antigène:UGT1A4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:54657
- Synonymes antigène:UGT1.4|UGT1A4|UDPGT 1-4|UGT1-04|UGT-1D|HUG-BR2|UGT1D|UDPGT
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Température de stockage:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 29-150 of human UGT1A4 (NP_009051.1).
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: UGT1A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse