Anti-KIT Rabbit Polyclonal Antibody
Catalog # PRSI60-079
Supplier: ProSci Inc.
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Mast/stem cell growth factor receptor Kit
- Symbole de l'antigène:KIT
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:937 - 966 aa
- Formulaire:Liquid
- ID de gène:3815
- Modification:None
- Molecular weight:110 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,32 mg/ml
- Température de transport:4°C
- Immunogène:This KIT antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 937 - 966 amino acids from the C-terminal region of human KIT.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Cdt:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 937-966
Host: Rabbit
Isotype: Ig
Reactivity: Human