Anti-GOLGA8H Rabbit Polyclonal Antibody
PRSI60-006
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Golgin subfamily A member 8H
- Symbole de l'antigène:GOLGA8H
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:19 - 45 aa
- Formulaire:Liquid
- ID de gène:728498
- Modification:None
- Molecular weight:71 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Température de transport:4°C
- Immunogène:This GOLGA8H antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 19 - 45 amino acids from the N-terminal region of human GOLGA8H.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Cdt:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GOLGA8H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 19-45
Host: Rabbit
Isotype: Ig
Reactivity: Human