Anti-TCHP Rabbit Polyclonal Antibody
Catalog # PRSI63-907
Supplier: ProSci Inc.
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Trichoplein keratin filament-binding protein
- Symbole de l'antigène:TCHP
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:291-320 aa
- Formulaire:Liquid
- ID de gène:84260
- Modification:None
- Molecular weight:61 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Température de transport:4 °C
- Immunogène:This TCHP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 291-320 amino acids from the Central region of human TCHP.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cdt:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: TCHP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 291-320
Host: Rabbit
Isotype: Ig
Reactivity: Human