Anti-CYP2C9 Rabbit Polyclonal Antibody
PRSI63-259
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Cytochrome P450 2C9
- Symbole de l'antigène:CYP2C9
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:82-110 aa
- Formulaire:Liquid
- ID de gène:1559
- Modification:None
- Molecular weight:56 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Température de transport:4 °C
- Immunogène:This CYP2C9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 82-110 amino acids from the N-terminal region of human CYP2C9.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cdt:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: CYP2C9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 82-110
Host: Rabbit
Isotype: Ig
Reactivity: Human