Anti-SULT1C2 Rabbit Polyclonal Antibody
Catalog # PRSI61-424
Supplier: ProSci Inc.
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Sulfotransferase 1C2
- Symbole de l'antigène:SULT1C2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:95 - 127 aa
- Formulaire:Liquid
- ID de gène:6819
- Modification:None
- Molecular weight:35 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Température de transport:4 °C
- Immunogène:This SULT1C1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 95 - 127 amino acids from the Central region of human SULT1C1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Cdt:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: SULT1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 95-127
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse