Anti-ARFGAP1 Rabbit Polyclonal Antibody
N° de catalogue PRSI61-363
Fournisseur: ProSci Inc.
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:ADP-ribosylation factor GTPase-activating protein 1
- Symbole de l'antigène:ARFGAP1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:377 - 406 aa
- Formulaire:Liquid
- ID de gène:55738
- Modification:None
- Molecular weight:45 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Température de transport:4 °C
- Immunogène:This ARFGAP1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 377 - 406 amino acids from the C-terminal region of human ARFGAP1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Cdt:400 µl
Spécifications
À propos de cet article
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 377-406
Host: Rabbit
Isotype: Ig
Reactivity: Human