Anti-POLR2H Rabbit Polyclonal Antibody
Catalog # PRSI60-956
Supplier: ProSci Inc.
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:DNA-directed RNA polymerases I, II, and III subunit RPABC3
- Symbole de l'antigène:POLR2H
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:59 - 87 aa
- Formulaire:Liquid
- ID de gène:5437
- Modification:None
- Molecular weight:17 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Température de transport:4 °C
- Immunogène:This POLR2H antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 59 - 87 amino acids from the Central region of human POLR2H.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cdt:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: POLR2H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 59-87
Host: Rabbit
Isotype: Ig
Reactivity: Human