Anti-VBP1 Rabbit Polyclonal Antibody
PRSI23-769
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:VHL binding protein 1
- Symbole de l'antigène:VBP1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:7411
- Synonymes antigène:VBP-1|von Hippel-Lindau binding protein 1|prefoldin subunit 3|PFDN3|HIBBJ46|PFD3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human VBP1 (NP_003363.1)
- Purification:Affinity purification.
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: VBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse