Anti-PTCD3 Rabbit Polyclonal Antibody
Catalog # PRSI23-625
Supplier: ProSci Inc.
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:pentatricopeptide repeat domain 3
- Symbole de l'antigène:PTCD3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:55037
- Synonymes antigène:pentatricopeptide repeat-containing protein 3|mitochondrial small ribosomal subunit protein mS39|28S ribosomal protein S39|TRG-15|mitochondrial|MRP-S39|transformation-related gene 15 protein|pentatricopeptide repeat domain-containing protein 3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Température de stockage:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 450-689 of human PTCD3 (NP_060422.4)
- Purification:Affinity purification.
- Type de conditionnement:Vial
- Cdt:50 µl
Specifications
About this item
Type: Primary
Antigen: PTCD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat