- Type d'anticorps:Primaire
- Nom de l'antigène:EF-CBP2
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:301-386/386
- Formulaire:Liquid
- ID de gène:871
- Synonymes antigène:NECA2_HUMAN|Stip 2|Synaptotagmin-interacting protein 2|Stip-2|N-terminal EF-hand calcium-binding protein 2|Neuronal calcium-binding protein 2|Necab2|Stip2|Synaptotagmin interacting protein 2|Neuronal calcium binding protein 2|EF hand calcium binding protein 2|EF-hand calcium-binding protein 2|EFCBP2|EF CBP2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human EF-CBP2
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Members of the EF-CBP (N-terminal EF-hand calcium binding protein)/NECAB (neuronal calcium-binding protein) family participate in neuronal calcium signaling. EF-CBP2, also known as NECAB2 (N-terminal EF-hand calcium binding protein 2), neuronal calcium-binding protein 2 or synaptotagmin-interacting protein 2 (Stip-2), is a 386 amino acid cytoplasmic protein that contains one antibiotic biosynthesis monooxygenase (ABM) domain and two EF-hand domains. Expressed in brain, EF-CBP2 is suggested to bind metabotropic glutamate receptor 5 (mGluR-5) in a calcium-regulated manner. The gene encoding EF-CBP2 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
Type: Primary
Antigen: EF-CBP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-386/386
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat