- Type d'anticorps:Primaire
- Nom de l'antigène:UBE2Q1
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:321-422/422
- Formulaire:Liquid
- ID de gène:55585
- Synonymes antigène:Ubiquitin carrier protein Q1|PRO3094|NICE5|Ubiquitin-conjugating enzyme E2 Q1|Ubiquitin conjugating enzyme E2 Q1|UBE2Q|GTAP|Galactosyl transferase associated protein|NICE 5|Protein NICE 5|Ubiquitin conjugating enzyme E2Q 1|Ubiquitin conjugating enzyme E2Q putative 1|Ubiquitin protein ligase Q1|Ube2q1|UB2Q1_HUMAN|Ubiquitin-protein ligase Q1|Protein NICE-5|Ubiquitin conjugating enzyme E2Q family member 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:46 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human UBE2Q1
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (UBE1s), ubiquitin-conjugating enzymes (UBE2s), and ubiquitin-protein ligases (UBE3s). When ubiquitin is activated by a UBE1, it is transferred to the cysteine residue on a UBE2. UBE2 then binds a UBE3, which transfers the ubiquitin from the UBE2 cysteine to a lysine residue on the target protein. Ubiquitin-conjugating enzyme E2 Q1 (UBE2Q1), also known as ubiquitin-protein ligase Q1 or ubiquitin carrier protein Q1, is an 422 amino acid protein belonging to the ubiquitin-conjugating enzyme (UBE2) family. Two named isoforms of UBE2Q1 exist as a result of alternative splicing events.
Type: Primary
Antigen: UBE2Q1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-422/422
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat