Anti-CEAM3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-6804R-A680
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:CEAM3
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:41-150/252
- Formulaire:Liquid
- ID de gène:1084
- Synonymes antigène:Carcinoembryonic antigen CGM1|carcinoembryonic antigen related cell adhesion molecule 3|carcinoembryonic|CEA|CEAM3_HUMAN|CEACAM 3|CEACAM-3|CGM1|CEAM-3|Ceacam3|W282|CD66d|W264|CEAM3|Carcinoembryonic antigen-related cell adhesion molecule 3|cd66 d|CEAM 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:23 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human CD66d/CEAM3
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Type: Primary
Antigen: CEAM3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-150/252
Host: Rabbit
Isotype: IgG
Reactivity: