Order Entry
Belgium
ContactUsLinkComponent
 
Anti-TIMM17A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6739R-A750
 :  Bioss
Anti-TIMM17A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6739R-A750
 :  Bioss

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    TIMM17A
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Alexa Fluor® 750
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    31-80/171
  • Formulaire:
    Liquid
  • ID de gène:
    10440
  • Synonymes antigène:
    mitochondrial inner membrane translocase|TIMM17A|preprotein translocase|Mitochondrial import inner membrane translocase subunit Tim17-A|Inner membrane preprotein translocase Tim17a|TI17A_HUMAN|TIM17A|TIM17|translocase of inner mitochondrial membrane 17 homolog A yeast
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    18 kDa
  • Température de stockage:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Température de transport:
    4 °C
  • Immunogène:
    KLH conjugated synthetic peptide derived from human TIMM17A
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Type de conditionnement:
    Vial
  • Cdt:
    100 µl

 

 

Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.Belongs to the Tim17/Tim22/Tim23 family.

Type: Primary
Antigen: TIMM17A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-80/171
Host: Rabbit
Isotype: IgG
Reactivity: