Anti-CKMT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6522R-A750
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:CKMT
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 750
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:115-220/417
- Formulaire:Liquid
- ID de gène:1159
- Synonymes antigène:Acidic-type mitochondrial creatine kinase|Mia-CK|CKMT1A|U-MtCK|CKMT|Ubiquitous mitochondrial creatine kinase|Creatine kinase U-type, mitochondrial|UMTCK|CKMT1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human Mitochondrial Creatine Kinase
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Reversibly catalyzes the transfer of phosphate between ATP and various phosphogens (e.g. creatine phosphate). Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.
Type: Primary
Antigen: CKMT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 115-220/417
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat