Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Estrogen Related Receptor gamma
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:61-160/458
- Formulaire:Liquid
- ID de gène:2104
- Synonymes antigène:ERR gamma-2|NR3B3|ERRG 2|ERR3|ESRRG|ERR3_HUMAN|ERRG2|ERR G2|Estrogen receptor-related protein 3|Estrogen receptor related protein 3|Estrogen-related receptor gamma|Nuclear receptor subfamily 3 group B member 3|ERR gamma 2|ERR 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:51 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human ESRRG/Estrogen Related Receptor gamma
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
This gene encodes a member of the estrogen receptor-related receptor (ESRR) family, which belongs to the nuclear hormone receptor superfamily. All members of the ESRR family share an almost identical DNA binding domain, which is composed of two C4-type zinc finger motifs. The ESRR members are orphan nuclear receptors; they bind to the estrogen response element and steroidogenic factor 1 response element, and activate genes controlled by both response elements in the absence of any ligands. The ESRR family is closely related to the estrogen receptor (ER) family. They share target genes, co-regulators and promoters, and by targeting the same set of genes, the ESRRs seem to interfere with the ER-mediated estrogen response in various ways. It has been reported that the family member encoded by this gene functions as a transcriptional activator of DNA cytosine-5-methyltransferases 1 (Dnmt1) expression by direct binding to its response elements in the DNMT1 promoters, modulates cell proliferation and estrogen signaling in breast cancer, and negatively regulates bone morphogenetic protein 2-induced osteoblast differentiation and bone formation. Multiple alternatively spliced transcript variants have been identified, which mainly differ at the 5' end and some of which encode protein isoforms differing in the N-terminal region.
Type: Primary
Antigen: Estrogen Related Receptor gamma
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/458
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat