Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CYP2A/Cytochrome P450 2A6
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:388-494/494
- Formulaire:Liquid
- ID de gène:1548
- Synonymes antigène:Cytochrome P450, family 2 subfamily A polypeptide 6|P450C2A|Cytochrome P450 subfamily IIA phenobarbital inducible polypeptide 6|CPA6|CYPIIA6|CYP2A6|P450PB|CP2A6_HUMAN|Flavoprotein linked monooxygenase|Cytochrome P450 2A6|Xenobiotic monooxygenase|Coumarin 7 hydroxylase|CYP2A3|Cytochrome P450I|Cytochrome P450 IIA3|CYP2A|Coumarin 7-hydroxylase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human CYP2A/Cytochrome P450 2A6
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
P450 enzymes constitute a family of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. Several P450 enzymes have been classified by sequence similarities as members of the CYP1A and CYP2A subfamilies. NADPH cytochrome P450 reductase is a microsomal enzyme responsible for the transfer of electrons from NADPH to cytochrome P450 enzymes during the P450 catalytic cycle. NADPH cytochrome P450 reductase is localised to the endoplasmic reticulum where it is also able to transfer electrons to heme oxygenase and cytochrome b5. NADPH cytochrome P450 reductase is structurally related to two separate flavoprotein families, ferredoxin nucleotide reductase (FNR) and flavodoxin. Electron transfer of NADPH cytochrome P450 reductase requires the binding of two flavin cofactors, FAD and FMN, to the FNR and flavodoxin domains, respectively.
Type: Primary
Antigen: CYP2A/Cytochrome P450 2A6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 388-494/494
Host: Rabbit
Isotype: IgG
Reactivity: