Anti-DZIP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-12191R-A680
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:DZIP3
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Mouse
- Environmentally Preferable:
- Épitope:245-350/1208
- Formulaire:Liquid
- ID de gène:9666
- Synonymes antigène:DZIP3_HUMAN|KIAA0675|DAZ-interacting protein 3|E3 ubiquitin-protein ligase DZIP3|UURF2|DAZ interacting protein 3, zinc finger|hRUL138|RNA-binding ubiquitin ligase of 138 kDa
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:139 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human DZIP3
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
DZIP3 is a 1208 amino acid protein that localizes to the cytoplasm and contains one RING-type zinc finger. Expressed in a variety of tissues with highest expression in heart, skeletal muscle and kidney, DZIP3 functions as an E3 ubiquitin-protein ligase that accepts ubiquitin from an E2 ubiquitin-conjugating enzyme, thereby playing a role in Signalling events throughout the cell. Multiple isoforms of DZIP3 exist due to alternative splicing events.
Type: Primary
Antigen: DZIP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 245-350/1208
Host: Rabbit
Isotype: IgG
Reactivity: Mouse