Anti-CABP5/CABP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12160R-A750
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CABP5/CABP3
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 750
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:21-120/173
- Formulaire:Liquid
- Synonymes antigène:Calcium binding protein 3|CaBP5|CABP5_HUMAN|Calcium-binding protein 5|CABP3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human CABP5/CABP3
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Specifications
About this item
The product of this gene belongs to a subfamily of calcium binding proteins, which share similarity to calmodulin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Expression of this gene is retina-specific. The mouse homolog of this protein has been shown to express in the inner nuclear layer of the retina, suggested its role in neuronal functioning. The specific function of this gene is unknown.
Type: Primary
Antigen: CABP5/CABP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/173
Host: Rabbit
Isotype: IgG
Reactivity: