Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:KIRREL3
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Mouse
- Environmentally Preferable:
- Épitope:351-450/778
- Formulaire:Liquid
- Synonymes antigène:Kin of IRRE like 3 Drosophila|KIRRE|PRO4502|KIRREL-3|yUNQ5923|KIRR3_HUMAN|KIRREL 3|MRD4|Nephrin like 2|Kin of IRRE like 3|NEPH2|PRO19814|Kin of irregular chiasm like protein 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:83 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human KIRREL3
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Specifications
About this item
NEPH2 is a 778 amino acid single-pass type I membrane protein that belongs to the nephrin-like protein family and immunoglobulin superfamily. Expressed in both fetal and adult brain, as well as podocytes of kidney glomeruli, NEPH2 contains five Ig-like C2-type (immunoglobulin-like) domains and is thought to play a role in the hematopoetic supportive capacity of stroma cells. NEPH2 undergoes alternative splicing to produce two isoforms and contains a C-terminal cytoplasmic domain which it uses to interact with Podocin, a podocyte protein involved in ultrafiltration. Defects in the gene encoding NEPH2 are associated with mental retardation autosomal dominant type 4 (MRD4).
Type: Primary
Antigen: KIRREL3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-450/778
Host: Rabbit
Isotype: IgG
Reactivity: Mouse