Anti-NPEPPS/PSA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
N° de catalogue BOSSBS-11684R-A680
Fournisseur: Bioss
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:NPEPPS/PSA
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Mouse
- Environmentally Preferable:
- Épitope:201-300/919
- Formulaire:Liquid
- Synonymes antigène:PSA|METALLOPROTEASE MP100|aminopeptidase puromycin sensitive|PSA_HUMAN|MP100|Puromycin sensitive aminopeptidase|Cytosol alanyl aminopeptidase|Npepps|Puromycin-sensitive aminopeptidase|AAP-S|MP 100|METALLOPROTEASE 100
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:103 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human NPEPPS
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
Puromycin-sensitive aminopeptidase is a 100 kDa zinc metallopeptidase which degrades neuropeptides by removing amino acid residues from the amino-terminus. The protein is the most abundant aminopeptidase in the brain, however it is not exclusive to that organ. It is localised primarily in the cytoplasm, and plays a role in the metabolism of neuropeptides in nerve terminals and synaptic clefts. The human PSA gene maps to chromosome 17q 2-32.
Type: Primary
Antigen: NPEPPS/PSA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/919
Host: Rabbit
Isotype: IgG
Reactivity: Mouse