Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CRSP9
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:101-200/233
- Formulaire:Liquid
- ID de gène:9443
- Synonymes antigène:MED7|Transcriptional coactivator CRSP33|Mediator complex subunit 7|MGC12284|RNA polymerase transcriptional regulation mediator subunit 7 homolog|CRSP33|ARC34|Activator-recruited cofactor 34 kDa component|MED7_HUMAN|CRSP complex subunit 9|hMED7|Cofactor required for Sp1 transcriptional activation subunit 9|Mediator of RNA polymerase II transcription subunit 7|Activator recruited cofactor 34 kDa component
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:27 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human CRSP9
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
In mammalian cells, transcription is regulated in part by high molecular weight coactivating complexes that mediate signals between transcriptional activators and RNA polymerase. These complexes include the SMCC (SRB and MED protein cofactor complex), which consists of various subunits that share homology with several components of the yeast transcriptional mediator complexes, and including the human proteins Srb7, Med6 (also designated DRIP33) and Med7 (also designated DRIP34). SMCC associates with the RNAPII (RNA polymerase II) holoenzyme through Srb7 and, in turn, enhances gene-specific activation or repression induced by DNA-binding transcription factors. Med6 and Med7, as well as other components of SMCC, associate with coactivator proteins from the TRAP (thyroid hormone receptor-activating protein) complex and DRIP (for vitamin D receptor interacting protein) complex to facilitate steroid receptor dependent transcriptional activation. Additionally, SMCC associates with PC4 (positive cofactor 4) to repress basal transcription independent of RNAPII activity.
Type: Primary
Antigen: CRSP9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/233
Host: Rabbit
Isotype: IgG
Reactivity: