Anti-CD64/IGFR1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-3511R-A680
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:CD64/IGFR1
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 680
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:101-200/374
- Formulaire:Liquid
- ID de gène:2209
- Synonymes antigène:FCGR1|FcgammaRIa|IgG Fc receptor I|Fc-gamma RI|FCG1|FCRI|FCGR1A|High affinity immunoglobulin gamma Fc receptor I|IGFR1|CD64|CD64A|Fc-gamma RIA
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:41 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human CD64/IGFR1
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
High affinity receptor for the Fc region of immunoglobulins gamma. Functions in both innate and adaptive immune responses.
Type: Primary
Antigen: CD64/IGFR1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/374
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat