Anti-PCSK4 Rabbit Polyclonal Antibody (Cy5)
BOSSBS-15501R-CY5
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:PCSK4
- Clonalité:Polyclonal
- Conjugaison:Cy5®
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:221-320/755
- Formulaire:Liquid
- ID de gène:54760
- Synonymes antigène:Proprotein convertase PC4|Proprotein convertase subtilisin/kexin type 4|PC4|Proprotein convertase 4|PCSK4_HUMAN|PCSK4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:70 kDa
- Température de stockage:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human PCSK4
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
PCSK4 is a member of the subtilisin-like proprotein convertase family. These enzymes process latent precursor proteins into their biologically active products. The encoded protein plays a critical role in reproduction and processes multiple prohormones including pro-pituitary adenylate cyclase-activating protein (proPACAP) and pro-insulin-like growth factor II.
Type: Primary
Antigen: PCSK4
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 221-320/755
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse