Anti-HEATR1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
BOSSBS-15438R-A750
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:HEATR1
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 750
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Mouse
- Environmentally Preferable:
- Épitope:51-150/2144
- Formulaire:Liquid
- ID de gène:55127
- Synonymes antigène:HEATR1|Protein BAP28|BAP28|HEAT repeat-containing protein 1|UTP10
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:242 kDa
- Température de stockage:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human HEATR1
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Involved in nucleolar processing of pre-18S ribosomal RNA. Involved in ribosome biosynthesis (By similarity).
Type: Primary
Antigen: HEATR1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/2144
Host: Rabbit
Isotype: IgG
Reactivity: Mouse